Skip to Content
Merck
All Photos(1)

Key Documents

92293

Sigma-Aldrich

Blue Dextran

average mol wt 40,000

Synonym(s):

Blue Dextran 40, Dextran blue from Leuconostoc mesenteroides

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352108

form

powder

mol wt

average mol wt 40,000

extent of labeling

0.01-0.08 mmol/g loading

loss

≤10% loss on drying

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

A fluorescent derivative of a dextran fraction with an average molecular weight of approximately 40000. It is supplied as a blue powder which is slowly soluble in water.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service