Skip to Content
Merck
All Photos(1)

Key Documents

92228

Sigma-Aldrich

D-Arabino-1,4-lactone

≥95.0% (GC)

Synonym(s):

D-Arabinonic acid γ-lactone, D-Arabinono-1,4-lactone, D-Arabonic acid-1,4-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O5
CAS Number:
Molecular Weight:
148.11
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95.0% (GC)

form

crystals

optical activity

[α]/D 72.0±8.0°, c = 1 in H2O

suitability

conforms to structure for Proton NMR spectrum

SMILES string

OC[C@H]1OC(=O)[C@@H](O)[C@@H]1O

InChI

1S/C5H8O5/c6-1-2-3(7)4(8)5(9)10-2/h2-4,6-8H,1H2/t2-,3-,4+/m1/s1

InChI key

CUOKHACJLGPRHD-JJYYJPOSSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neha Biyani et al.
Molecular and biochemical parasitology, 180(2), 76-85 (2011-09-13)
Plants and most animals can synthesize ascorbate (vitamin C) for their own requirements, but humans have lost this ability during evolution. The last step in the biosynthesis of L-ascorbic acid involves the conversion of an aldonolactone substrate to ascorbate (or
Benjamin Metz et al.
Eukaryotic cell, 10(11), 1527-1535 (2011-09-06)
The conidium plays a critical role in the life cycle of many filamentous fungi, being the primary means for survival under unfavorable conditions. To investigate the transcriptional changes taking place during the transition from growing hyphae to conidia in Trichoderma
Reza Khorassani et al.
BMC plant biology, 11, 121-121 (2011-08-30)
In soils with a low phosphorus (P) supply, sugar beet is known to intake more P than other species such as maize, wheat, or groundnut. We hypothesized that organic compounds exuded by sugar beet roots solubilize soil P and that
Merlin Eric Hobbs et al.
Biochemistry, 52(1), 239-253 (2012-12-12)
A member of the amidohydrolase superfamily, BmulJ_04915 from Burkholderia multivorans, of unknown function was determined to hydrolyze a series of sugar lactones: L-fucono-1,4-lactone, D-arabino-1,4-lactone, L-xylono-1,4-lactone, D-lyxono-1,4-lactone, and L-galactono-1,4-lactone. The highest activity was shown for L-fucono-1,4-lactone with a k(cat) value of
Marta Gutierrez-Larrainzar et al.
Journal of agricultural and food chemistry, 58(19), 10631-10638 (2010-09-08)
D-erythroascorbate (D-EAA), a five-carbon analogue of L-ascorbate (L-AA), and D-erythroascorbate monoglucoside (D-EAAG) are accumulated in Phycomyces blakesleeanus grown on glucose (99.5 and 1084 μg/g mycelial dry weight, respectively) and also excreted into the culture medium. Both compounds showed UV spectral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service