Skip to Content
Merck
All Photos(1)

Key Documents

90421C

Sigma-Aldrich

Sodium bicarbonate

dry powder, suitable for cell culture, suitable for insect cell culture

Synonym(s):

Sodium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHCO3
CAS Number:
Molecular Weight:
84.01
Beilstein:
4153970
EC Number:
MDL number:
UNSPSC Code:
12352200

description

for research or for further manufacturing use

form

dry powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

density

2.16 g/mL at 25 °C (lit.)

SMILES string

[Na+].OC([O-])=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

UIIMBOGNXHQVGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Athanasia Petala et al.
Water research, 81, 157-166 (2015-06-10)
Ethyl paraben (EP), an emerging micro-pollutant representative of the parabens family, has been subject to photocatalytic degradation under simulated solar radiation at a photon flux of 1.3·10(-4) E/(m(2) s). Six nitrogen-doped titania catalysts synthesized by annealing a sol-gel derived TiO2 powder under
Alessio Innocenti et al.
Bioorganic & medicinal chemistry letters, 15(3), 573-578 (2005-01-25)
A detailed inhibition study of five carbonic anhydrase (CA, EC 4.2.1.1) isozymes with carboxylates including aliphatic (formate, acetate), dicarboxylic (oxalate, malonate), hydroxy/keto acids (l-lactate, l-malate, pyruvate), tricarboxylic (citrate), or aromatic (benzoate, tetrafluorobenzoate) representatives, some of which are important intermediates in
Alessandra Cincinelli et al.
Bioorganic & medicinal chemistry, 19(6), 1847-1851 (2011-03-08)
A high activity α-carbonic anhydrase (CA, EC 4.2.1.1) has been purified from various tissues of the Antarctic seal Leptonychotes weddellii. The new enzyme, denominated lwCA, has a catalytic activity for the physiologic CO(2) hydration to bicarbonate reaction, similar to that
Peter Burghout et al.
Bioorganic & medicinal chemistry, 19(1), 243-248 (2010-12-18)
The Gram-positive bacterium Streptococcus pneumoniae is a human respiratory tract pathogen that contributes significantly to global mortality and morbidity. It was recently shown that this bacterial pathogen depends on a conserved β-carbonic anhydrase (CA, EC 4.2.1.1) for in vitro growth
Daniela Vullo et al.
Bioorganic & medicinal chemistry letters, 21(12), 3591-3595 (2011-05-17)
Two new β-carbonic anhydrases (CAs, EC 4.2.1.1) from the bacterial pathogen Salmonella enterica serovar Typhimurium, stCA 1 and stCA 2, were characterized kinetically. The two enzymes possess appreciable activity as catalysts for the hydration of CO(2) to bicarbonate, with k(cat)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service