Skip to Content
Merck
All Photos(1)

Key Documents

89283

Sigma-Aldrich

4-(2-Methyl-4-thiazolyl)phenol

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NOS
CAS Number:
Molecular Weight:
191.25
Beilstein:
1104995
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98.0% (HPLC)

form

crystals

SMILES string

Cc1nc(cs1)-c2ccc(O)cc2

InChI

1S/C10H9NOS/c1-7-11-10(6-13-7)8-2-4-9(12)5-3-8/h2-6,12H,1H3

InChI key

HOAYTTLLVORNLA-UHFFFAOYSA-N

Application

suitable as enhancer for luminol chemiluminescence

Other Notes

Potent peroxidase chemiluminescence enhancer for HRP

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service