Skip to Content
Merck
All Photos(1)

Key Documents

83219

Millipore

Protein G

immobilized on cross-linked 4% agarose

Synonym(s):

Protein G agarose

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$1,012.00
250 MG
R$2,022.00
1 G
R$6,318.00

R$1,012.00


Estimated to ship onMay 07, 2025



Select a Size

Change View
100 MG
R$1,012.00
250 MG
R$2,022.00
1 G
R$6,318.00

About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

R$1,012.00


Estimated to ship onMay 07, 2025


Product Name

Protein G, immobilized on cross-linked 4% agarose,

form

powder

capacity

≥15 mg/mL, gel binding capacity (human IgM)

storage temp.

−20°C

General description

protein G, attached to 4% cross-linked beaded agarose, stabilized with lactose which must be removed prior to use by washing gel onto filter; 1 g powder yields ~4 mL swollen gel

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Liu et al.
Genome research, 15(3), 421-427 (2005-02-16)
We have developed a new method for determining the DNA-binding specificity of proteins. In DIP-chip (DNA immunoprecipitation with microarray detection), protein.DNA complexes are isolated from an in vitro mixture of purified protein and naked genomic DNA. Whole-genome DNA microarrays are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service