Skip to Content
Merck
All Photos(1)

Key Documents

78787

Supelco

Phenyl isothiocyanate solution

for protein sequence analysis, ~5% in heptane

Synonym(s):

PITC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NCS
CAS Number:
Molecular Weight:
135.19
Beilstein:
471392
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.22

grade

for protein sequence analysis

form

solution

concentration

~5% in heptane

technique(s)

HPLC: suitable

suitability

in accordance for sequence analysis test

storage temp.

−20°C

SMILES string

S=C=Nc1ccccc1

InChI

1S/C7H5NS/c9-6-8-7-4-2-1-3-5-7/h1-5H

InChI key

QKFJKGMPGYROCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Special packaging (40 ml) for direct use in common sequencers.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yungang Lan et al.
Molecular neurobiology, 57(12), 5299-5306 (2020-09-03)
Lysosomes are involved in pathogenesis of a variety of neurodegenerative diseases and play a large role in neurodegenerative disorders caused by virus infection. However, whether virus-infected cells or animals can be used as experimental models of neurodegeneration in humans based
R Gheshlaghi et al.
Analytical biochemistry, 383(1), 93-102 (2008-08-30)
Modified resolution and overall separation factors used to quantify the separation of complex chromatography systems are described. These factors were proven to be applicable to the optimization of amino acid resolution in reverse-phase (RP) HPLC chromatograms. To optimize precolumn derivatization
Fantao Meng et al.
Bioconjugate chemistry, 19(7), 1352-1360 (2008-06-25)
Successful and cost-effective PEGylation protocols require pure functionalized PEG reagents, which can be synthesized by simple and efficient procedures, exhibit high stability against hydrolysis, and maintain a level of reactivity with protein functional groups under mild reaction conditions. PEG-phenyl-isothiocyanate (PIT-PEG)
Misako Aito-Inoue et al.
Journal of agricultural and food chemistry, 54(15), 5261-5266 (2006-07-20)
For the isolation and detection of food-derived peptides in blood, an approach based on the derivatization of peptides with phenyl isothiocyanate (PITC) was developed. This approach allows hydrophilic peptides to be resolved and specifically detected by reversed-phase (RP) HPLC. For
R Thippeswamy et al.
Journal of agricultural and food chemistry, 54(19), 7014-7019 (2006-09-14)
Two liquid chromatographic methods that involve precolumn derivatization with o-phthaladehyde (OPA) and phenylisothiocyanate (PITC) with fluorescence and diode array UV detection for the determination of theanine have been developed. The chromatographic separations were achieved by reverse-phase high-performance liquid chromatography using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service