Skip to Content
Merck
All Photos(1)

Key Documents

74576

Sigma-Aldrich

1,2-Dierucoyl-sn-glycero-3-phosphoethanolamine

≥98% (TLC)

Synonym(s):

1,2-Di(cis-13-docosenoyl)-sn-glycero-3-phosphoethanolamine, DEPE, Dierucoyl phosphatidylethanolamine, PE(22:1(13E)/22:1(13E))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H94NO8P
CAS Number:
Molecular Weight:
856.25
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.85

Assay

≥98% (TLC)
≥98% (as erucic acid)

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)OCCN)OC(=O)CCCCCCCCCCC\C=C/CCCCCCCC

InChI

1S/C49H94NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-41-48(51)55-45-47(46-57-59(53,54)56-44-43-50)58-49(52)42-40-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,47H,3-16,21-46,50H2,1-2H3,(H,53,54)/b19-17-,20-18-/t47-/m1/s1

InChI key

VBZSMBBOZFITID-FRWASNMLSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

>392.0 °F

Flash Point(C)

> 200 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boris Tenchov et al.
Chemistry and physics of lipids, 208, 65-74 (2017-10-07)
The non-lamellar phases formed by membrane lipids in diluted aqueous dispersions are mainly represented by the inverted hexagonal phase, HII, and phases of cubic symmetry, among them the bicontinuous cubic phases Pn3m (Q224), Im3m (Q229) and Ia3d (Q230). Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service