Skip to Content
Merck
All Photos(1)

Documents

72543

Sigma-Aldrich

Nitr 5/AM

suitable for fluorescence, ≥90% (HPLC)

Synonym(s):

Nitr 5 tetrakis(acetoxymethyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H47N3O23
CAS Number:
Molecular Weight:
973.84
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥90% (HPLC)

form

solid

suitability

suitable for fluorescence

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)c1ccc(C)cc1OCCOc2cc(ccc2N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)C(O)c3cc4OCOc4cc3[N+]([O-])=O

InChI

1S/C43H47N3O23/c1-25-6-8-32(44(16-39(51)66-20-60-26(2)47)17-40(52)67-21-61-27(3)48)35(12-25)58-10-11-59-36-13-30(43(55)31-14-37-38(65-24-64-37)15-34(31)46(56)57)7-9-33(36)45(18-41(53)68-22-62-28(4)49)19-42(54)69-23-63-29(5)50/h6-9,12-15,43,55H,10-11,16-24H2,1-5H3

InChI key

KGQNBQFELVBWSX-UHFFFAOYSA-N

Application

Acetoxymethyl ester of Nitr 5. For loading the chelator into the cell where it is cleaved by cytoplasmic esterases.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service