Recommended Products
Assay
≥97.0% (HPLC)
optical activity
[α]/D -13.0±1.0°, c = 1 in H2O
storage temp.
−20°C
SMILES string
OC[C@@H]1OC(O)[C@@H](O)[C@@H](O)[C@H]1O
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4-,5-,6?/m0/s1
InChI key
WQZGKKKJIJFFOK-HOWGCPQDSA-N
Application
L-Allose is a pure enantiomer that may be used with other aldose substrates such as D-lyxose, D-mannose, L-ribose, and D-talose to identify, differentiate and characterize aldose isomerase(s) and epimerase(s).
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of organic chemistry, 73(14), 5636-5639 (2008-06-24)
An expeditious and efficient synthesis of 1,6-anhydro-beta-L-hexopyranosyl derivatives 3 as valuable building blocks for the preparation of L-sugars is herein reported. This route relies upon the use of a domino reaction involving five synthetic steps from the 5,6-dihydro-1,4-dithiin 4. As
Journal of bacteriology, 189(5), 1655-1663 (2006-12-26)
A newly isolated bacterium, Cohnella laevoribosii RI-39, could grow in a defined medium with L-ribose as the sole carbon source. A 21-kDa protein isomerizing L-ribose to L-ribulose, as well as D-lyxose to D-xylulose, was purified to homogeneity from this bacterium.
Analytical and bioanalytical chemistry, 401(2), 689-698 (2011-06-03)
D-Allose, a rare, naturally occurring monosaccharide, is known to exert anti-proliferative effects on cancer cells. The effects of D-allose on the cellular membranes of hormone-refractory prostate cancer cell line (DU145), hormone-sensitive prostate cancer cell line (LNCaP), and normal prostate epithelial
Journal of bioscience and bioengineering, 112(6), 638-642 (2011-09-06)
The anti-oxidative activity of the rare sugar D-allose has recently been reported, but the mechanism is largely unclear. In this study, we evaluated the reactive oxygen species (ROS) scavenging activities of D-allose and then examined the effects of D-allose on
Journal of bioscience and bioengineering, 110(1), 26-31 (2010-06-15)
The specific activity and catalytic efficiency (k(cat)/K(m)) of the recombinant putative protein from Providencia stuartii was the highest for D-lyxose among the aldose substrates, indicating that it is a D-lyxose isomerase. Gel filtration analysis suggested that the native enzyme is
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service