Skip to Content
Merck
All Photos(1)

Key Documents

59041

Sigma-Aldrich

2,2,4-Trimethylpentane

for luminescence, ≥99.5% (GC)

Synonym(s):

Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for luminescence

vapor density

3.9 (vs air)

vapor pressure

41 mmHg ( 21 °C)

Assay

≥99.5% (GC)

autoignition temp.

745 °F

expl. lim.

6 %

refractive index

n20/D 1.391 (lit.)
n20/D 1.391

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

density

0.692 g/mL at 25 °C (lit.)

λ

1 cm path, H2O reference

UV absorption

λ: 210 nm Amax: 0.5
λ: 220 nm Amax: 0.14
λ: 230 nm Amax: 0.06
λ: 240 nm Amax: 0.02
λ: 255 nm Amax: 0.005

suitability

in accordance for luminescence

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yufeng Zhang et al.
Journal of chromatography. A, 1252, 67-73 (2012-07-10)
In this study, a polyethylene Pasteur pipette-based ultrasound-assisted emulsification microextraction (USAEME) applying low-density organic solvent was successfully developed for the extraction of trace levels of organochlorine pesticides (OCPs) in water samples and followed by gas chromatography-mass spectrometry (GC-MS) analysis. In
Luciane Batistella et al.
Bioprocess and biosystems engineering, 35(3), 351-358 (2011-07-23)
Both stability and catalytic activity of two commercial immobilized lipases were investigated in the presence of different organic solvents in ultrasound-assisted system. In a general way, for Novozym 435, the use of ethanol as solvent led to a loss of
Hisham A El-Masri et al.
Inhalation toxicology, 21(14), 1176-1185 (2009-11-20)
2,2,4-Trimethylpentane (TMP) is a volatile colorless liquid used primarily to increase the octane rating of combustible fuels. TMP is released in the environment through the manufacture, use, and disposal of products associated with the gasoline and petroleum industry. Short-term inhalation
Kyung Min Park et al.
Food chemistry, 138(2-3), 733-738 (2013-02-16)
In this study, we investigated the possibility of reversed micelles, widely used as an enzyme reactor for lipases, for the determination of lipoxygenase activity. Although it is rapid and simple, reversed micelles have some limitations, such as interference by UV-absorbing
Aram Kang et al.
Molecular bioSystems, 8(4), 1350-1358 (2012-02-14)
Microbial tolerance to hydrocarbons has been studied in an effort to improve the productivity of biochemical processes and to enhance the efficiency of hydrocarbon bioremediation. Despite these studies, few attempts have been made to design rational strategies to improve microbial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service