Skip to Content
Merck
All Photos(1)

Key Documents

54008

Sigma-Aldrich

1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine

≥98.0% (TLC)

Synonym(s):

(9Z)-9-Octadecenoic acid (1R)-1-[[[(2-aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester, 1,2-Di(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine, 3-sn-Phosphatidylethanolamine, 1,2-dideoyl, L-β,γ-Dioleoyl-α-cephalin, DOPE, Dioleoyl phosphatidylethanolamine, PE(18:1(9Z)/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H78NO8P
CAS Number:
Molecular Weight:
744.03
Beilstein:
1718431
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥98.0% (TLC)

form

solid

functional group

ester

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OCC(COP(O)(=O)OCCN)OC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C41H78NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(43)47-37-39(38-49-51(45,46)48-36-35-42)50-41(44)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,39H,3-16,21-38,42H2,1-2H3,(H,45,46)/b19-17-,20-18-

InChI key

MWRBNPKJOOWZPW-CLFAGFIQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE) forms heterogenous liposomes with N-[1-(2,3-dioleoyloxy)propyl]-N,N,N-trimethylammonium methylsulfate (DOTAP), which are used as delivery vehicles for therapeutic agents .

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio A M Gasperini et al.
Langmuir : the ACS journal of surfaces and colloids, 31(11), 3308-3317 (2015-03-03)
This work presents a study of the association between low molecular weight hyaluronic acid (16 kDa HA) and cationic liposomes composed of egg phosphatidylcholine (EPC), 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE), and 1,2-dioleoyl-3-trimethylammonium-propane (DOTAP). The cationic liposome/HA complexes were evaluated to determine their mesoscopic
Jonathan Langdown et al.
Blood, 124(12), 1951-1956 (2014-07-23)
In this study, we describe a novel thrombomodulin (TM) mutation (c.1611C>A) that codes for a change from cysteine 537 to a premature stop codon (p.Cys537Stop). Three members of a family with a history of posttraumatic bleeding were identified to be
Ryo Furukawa et al.
Biomaterials, 57, 107-115 (2015-04-29)
Mitochondrial genome-targeting nucleic acids are promising therapeutic candidates for treating mitochondrial diseases. To date, a number of systems for delivering genetic information to the cytosol and the nucleus have been reported, and several successful gene therapies involving gene delivery targeted
S Peraramelli et al.
Journal of thrombosis and haemostasis : JTH, 12(11), 1826-1837 (2014-08-29)
TFPI is a Kunitz-type protease inhibitor that downregulates the extrinsic coagulation pathway by inhibiting factor Xa (FXa) and FVIIa. All three Kunitz domains (KD1, KD2, and KD3) and protein S are required for optimal inhibition of FXa and FVIIa. There
Muneeb Ahmed et al.
PloS one, 10(7), e0128910-e0128910 (2015-07-15)
Radiofrequency thermal ablation (RFA) of hepatic and renal tumors can be accompanied by non-desired tumorigenesis in residual, untreated tumor. Here, we studied the use of micelle-encapsulated siRNA to suppress IL-6-mediated local and systemic secondary effects of RFA. We compared standardized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service