Skip to Content
Merck
All Photos(1)

Key Documents

53264

Sigma-Aldrich

(R)-γ-Valerolactone

≥97.5% (GC)

Synonym(s):

(5R)-4-Hydroxypentanoic acid lactone, (5R)-5-Methyloxolan-2-one, (R)-γ-Methyl-γ-butyrolactone, (R)-5-Methyl-2-oxotetrahydrofuran, (R)-Dihydro-5-methyl-2(3H)-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
Beilstein:
3648300
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥97.5% (GC)

form

liquid

optical purity

enantiomeric excess: ≥96.5% (GC)

storage temp.

2-8°C

SMILES string

C[C@@H]1CCC(=O)O1

InChI

1S/C5H8O2/c1-4-2-3-5(6)7-4/h4H,2-3H2,1H3/t4-/m1/s1

InChI key

GAEKPEKOJKCEMS-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Metabolite for urine analysis, metabolite of interest in pathways of biomass utilization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gorissen, H. J., et al.
Chirality, 4, 286-294 (1992)
Shinji Nakamura et al.
Journal of the American Chemical Society, 126(28), 8769-8776 (2004-07-15)
The mechanism of the optical resolution of gamma-valerolactone (VAL) enantiomers by enclathration in cholic acid (CA) channels was investigated. 13C cross-polarization magic-angle spinning spectra of CA/VAL inclusion compounds show four methyl 13C peaks of VAL with different intensities depending on
Identification of the metabolites of n-hexane, cyclohexane, and their isomers in men's urine.
L Perbellini et al.
Toxicology and applied pharmacology, 53(2), 220-229 (1980-04-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service