Skip to Content
Merck
All Photos(1)

Documents

51282

Sigma-Aldrich

Deoxyfuconojirimycin hydrochloride

≥98.0% (TLC)

Synonym(s):

1,5-Dideoxy-1,5-imino-L-fucitol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO3 · HCl
CAS Number:
Molecular Weight:
183.63
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98.0% (TLC)

SMILES string

Cl.C[C@@H]1NC[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C6H13NO3.ClH/c1-3-5(9)6(10)4(8)2-7-3;/h3-10H,2H2,1H3;1H/t3-,4+,5+,6-;/m0./s1

InChI key

SHSBWMUIVLOMIL-NQZVPSPJSA-N

Biochem/physiol Actions

Deoxyfuconojirimycin is a specific and competitive inhibitor of human liver α-L-fucosidase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun Yuan et al.
Pathology oncology research : POR, 14(2), 145-156 (2008-06-17)
Glycosylation drives critical processes important for mammalian cell-cell and cell-matrix interactions. Alpha-L-fucose (alpha-L-f) is a key monosaccharide component of oligosaccharides that has been found to be overexpressed during tumor progression. Modification of cell surface fucosylation, we hypothesized, alters tumor cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service