Skip to Content
Merck
All Photos(1)

Key Documents

44700

Sigma-Aldrich

Octadecylphosphophosphoniumcholine

≥98.0% (TLC)

Synonym(s):

Octadecyl-[2-(trimethylphosphonio)-ethyl]-phosphat

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H50O4P2
CAS Number:
Molecular Weight:
452.59
Beilstein:
7143715
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.85

Assay

≥98.0% (TLC)

lipid type

phospholipids

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCOP([O-])(=O)OCC[P+](C)(C)C

InChI

1S/C23H50O4P2/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-26-29(24,25)27-22-23-28(2,3)4/h5-23H2,1-4H3

InChI key

LKRUUBNORFYVBG-UHFFFAOYSA-N

Application

Antitumor phospholipid, cytotoxic activity on carcinoma cell cultures

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Qin et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1124, 82-99 (2019-06-10)
The abuse of fentanyl and its analogues, which are potent, short-acting, synthetic narcotic analgesics, has become a major issue. Many cases containing fentanyl-analogue and novel synthetic opioids have also been reported. Hence, we report the determination of fentanyl, fentanyl-analogue and
Stekar, J., et al.
Angewandte Chemie (International Edition in English), 34, 238-240 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service