Skip to Content
Merck
All Photos(1)

Key Documents

39104

Sigma-Aldrich

Ethylene glycol disuccinate bis(sulfo-N-succinimidyl) ester sodium salt

puriss., suitable for protein modification, ≥95.0% (dry substance, CHN)

Synonym(s):

Bis(sulfo-N-succinimidyl) ethylene glycol disuccinate sodium salt, Ethylene glycol bis(sulfosuccinimidyl succinate) sodium salt, Sulfo-EGS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N2Na2O18S2
CAS Number:
Molecular Weight:
660.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

puriss.

Assay

≥95.0% (dry substance, CHN)

impurities

≤10% water

suitability

suitable for protein modification

storage temp.

−20°C

SMILES string

O=C(CC1S(=O)([O-])=O)N(OC(CCC(OCCOC(CCC(ON2C(CC(S(=O)([O-])=O)C2=O)=O)=O)=O)=O)=O)C1=O.[Na+].[Na+]

InChI

1S/C18H20N2O18S2.2Na/c21-11-7-9(39(29,30)31)17(27)19(11)37-15(25)3-1-13(23)35-5-6-36-14(24)2-4-16(26)38-20-12(22)8-10(18(20)28)40(32,33)34;;/h9-10H,1-8H2,(H,29,30,31)(H,32,33,34);;/q;2*+1/p-2

InChI key

IYBKWXQWKPSYDT-UHFFFAOYSA-L

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Water-soluble reagent for cross-linking

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Kuramitz et al.
Analytical Chemistry, 74, 553-553 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service