Skip to Content
Merck
All Photos(1)

Key Documents

32878

Sigma-Aldrich

Fuchsin solution

for microscopy, denatured with 2-butanone, 2% ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein:
4166684
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

for microscopy

quality

denatured with 2-butanone

concentration

2% ethanol

suitability

suitable for microscopy

SMILES string

Br.CO[C@@H]1CCCN[C@H]1CC(C)=O

InChI

1S/C9H17NO2.BrH/c1-7(11)6-8-9(12-2)4-3-5-10-8;/h8-10H,3-6H2,1-2H3;1H/t8-,9+;/m0./s1

InChI key

DVSQBUUDEVNABC-OULXEKPRSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service