Skip to Content
Merck
All Photos(1)

Documents

240R-1

Sigma-Aldrich

COX-2 (SP21) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP21, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (240R-14)
vial of 0.5 mL concentrate (240R-15)
bottle of 1.0 mL predilute (240R-17)
vial of 1.0 mL concentrate (240R-16)
bottle of 7.0 mL predilute (240R-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

colon carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... PTGS2(5743)

General description

Cyclooxygenase 2 (COX-2) catalyzes the conversion of arachidonic acid to prostaglandin H2 in the first step in the biosynthesis of prostaglandins, thromboxanes, and prostacyclins. COX-2 inhibition by nonsteroidal anti-inflammatory agents has been shown to decrease angiogenesis and tumor growth, and promote apoptosis. COX-2 overexpression has been associated with increased microvascular density.

Quality


IVD

IVD

IVD

RUO

Linkage

COX-2 Positive Control Slides, Product No. 240S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oreste Gallo et al.
Human pathology, 33(7), 708-714 (2002-08-28)
Prostaglandins play a critical role in tumor development and growth by regulating numerous biologic processes, including tumor angiogenesis, with clear prognostic and therapeutic implications. The aim of this study was to investigate the prognostic relevance of cyclooxygenase-2 (COX-2) pathway activation
H Sano et al.
Cancer research, 55(17), 3785-3789 (1995-09-01)
Several studies indicate that nonsteroidal anti-inflammatory drugs including indomethacin, aspirin, sulindac, and piroxicam reduce the risk of colon cancer. Furthermore, nonsteroidal anti-inflammatory drugs that inhibit the cyclooxygenase (COX) enzyme were shown to inhibit the development of colon cancer in animal
Jan Stoehlmacher et al.
Seminars in oncology, 30(3 Suppl 6), 10-16 (2003-06-13)
Cyclooxygenase (COX) enzyme-dependent arachidonic acid metabolites occupy key positions in important physiologic processes such as immunity, reproduction, and vascular integrity. Large retrospective and prospective population-based studies have shown that the use of both nonselective, nonsteroidal anti-inflammatory drugs and selective COX-2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service