Skip to Content
Merck
All Photos(1)

Key Documents

83353

Sigma-Aldrich

Mercury(II) bromide

puriss. p.a., ACS reagent, ≥99.0% (precipitation titration)

Synonym(s):

Mercuric bromide

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
R$861.00

R$861.00


Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
50 G
R$861.00

About This Item

Linear Formula:
HgBr2
CAS Number:
Molecular Weight:
360.40
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
form:
powder or crystals

R$861.00


Available to ship onApril 07, 2025Details


Request a Bulk Order

grade

ACS reagent
puriss. p.a.

vapor pressure

1 mmHg ( 136.5 °C)

Assay

≥99.0% (precipitation titration)

form

powder or crystals

ign. residue (after reduction)

≤0.02%

bp

322 °C (lit.)

mp

236 °C (lit.)

anion traces

chloride (Cl-): ≤2000 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

Br[Hg]Br

InChI

1S/2BrH.Hg/h2*1H;/q;;+2/p-2

InChI key

NGYIMTKLQULBOO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Mercury(II) bromide (HgBr2) affords 1:1 adducts on reaction with N,N,N′,N′- tetramethyl-o-phenylenediamine (o-Me2N.C6H4.NMe2). Single-crystal X-ray diffraction studies of these adducts have been reported.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal Structures of the 1: 1 Adducts of N, N, N', N'-Tetramethyl-o-phenylenediamine with Zinc (II) Bromide and Mercury (II) Bromide and Iodide.
Hughes CM, et al.
Australian Journal of Chemistry, 38(10), 1521-1527 (1985)
S Engst et al.
Biochemistry, 37(33), 11496-11507 (1998-08-26)
Conditions are described under which the nonphysiological substrate mercuric bromide (HgBr2) is rapidly turned over, both by the wild type (CCCC) and by an active site double mutant (CCAA) of mercuric reductase in which the C-terminal cysteines 557' and 558'
Don S Forsyth et al.
Food additives and contaminants, 21(9), 849-856 (2005-01-26)
Mercury was detected in all analysed samples of swordfish, marlin, shark and tuna purchased from major supermarket outlets and fish retailers in three cities across Canada. Total mercury and methylmercury levels ranged up to 3845 and 2346 ng g(-1), respectively.
M E Goodsite et al.
Environmental science & technology, 38(6), 1772-1776 (2004-04-13)
The oxidation of elemental mercury (Hg0) to the divalent gaseous mercury dibromide (HgBr2) has been proposed to account for the removal of Hg0 during depletion events in the springtime Arctic. The mechanism of this process is explored in this paper
G Medgyes et al.
Carbohydrate research, 186(2), 225-239 (1989-03-15)
D-Galactose was converted into the glycosylating agents 4-azido-2,3-di-O-benzyl-4-deoxy-6-O-propionyl-alpha-D-glucopyranosyl chloride (11) and the methyl beta-D-thiopyranoside 19. Condensation of 11 with 2,5-diazido-1,6-di-O-benzoyl-2,5-di-deoxy-L-iditol in the presence of mercury salts gave 24% of 2,5-diazido-3-O-(4-azido-2,3-di-O-benzyl-4-deoxy-6-O-propionyl-alp ha-D- glucopyranosyl)-1,6-di-O-benzoyl-2,5-dideoxy-L-iditol. Methyl trifluoromethanesulfonate-promoted glycosylation of 1,3-diazido-2-O-benzyl-1,3-dideoxy-5,6-O-isopropylidene-D-gulit ol with 19

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service