Skip to Content
Merck
All Photos(1)

Key Documents

72260

Sigma-Aldrich

Nickel(II) perchlorate hexahydrate

purum p.a., crystallized, ≥98.0% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(ClO4)2 · 6H2O
CAS Number:
Molecular Weight:
365.69
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
form:
crystals

grade

purum p.a.

Assay

≥98.0% (KT)

form

crystals

quality

crystallized

reaction suitability

reagent type: oxidant

mp

140 °C (lit.)

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤500 mg/kg
Cd: ≤50 mg/kg
Co: ≤500 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤200 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.O.O.O.[Ni++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.Ni.6H2O/c2*2-1(3,4)5;;;;;;;/h2*(H,2,3,4,5);;6*1H2/q;;+2;;;;;;/p-2

InChI key

WHGYCGOFTBFDLW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) perchlorate hexahydrate is a hydrated nickel(II) salt.

Application

Nickel(II) perchlorate hexahydrate may be used in the synthesis of tris(2,2′-bipyridylamine)nickel(II) perchlorate[1] and 4,7,7-trimethyl-3,4,5,6,7,8-hexahydrobenzopyran-2(H),5-dione.[2]

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 2 - Ox. Sol. 3 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel (II)-2, 2'-dipyridylamine system. I. Synthesis and stereochemistry of the complexes.
Hurley TJ and Robinson MA.
Inorganic Chemistry, 7(1), 33-38 (1968)
A new method for enol lactone synthesis by a Michael addition/cyclization sequence.
Itoh K and Kanemasa S.
Tetrahedron Letters, 44(9), 1799-1802 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service