Skip to Content
Merck
All Photos(1)

Key Documents

63090

Sigma-Aldrich

Magnesium oxide

puriss. p.a., ACS reagent, ≥97% (calcined substance, KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent
puriss. p.a.

Assay

≥97% (calcined substance, KT)

form

powder

impurities

≤0.002% total nitrogen (N)

loss

≤2% loss on ignition, 800°C

mp

2852 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate, sulfite (as SO42-): ≤200 mg/kg

cation traces

Ba: ≤10 mg/kg
Ca: ≤500 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mn: ≤5 mg/kg
Na: ≤2500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤20 mg/kg
Zn: ≤5 mg/kg

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium oxide (MgO) may be employed as a phase stabilizer during the fabrication of porous β-tricalcium phosphate (β-TCP).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eagleson M.
Concise Encyclopedia Chemistry, 495-495 (1994)
Magnesium oxide-catalysed reaction of carbon dioxide with an epoxide with retention of stereochemistry.
Yano T, et al.
Chemical Communications (Cambridge, England), 12, 1129-1130 (1997)
Synthesis of ethylene and ethane by partial oxidation of methane over lithium-doped magnesium oxide.
Ito T and Lunsford JH.
Nature, 721-722 (1985)
Fabrication of β-TCP foam: Effects of magnesium oxide as phase stabilizer on its properties.
Nikaido T, et al.
Ceramics International, 41(10), 14245-14250 (2015)
Qiulin Yang et al.
Bioresource technology, 123, 49-54 (2012-09-04)
The cooking with solid alkali and active oxygen has a high selectivity for delignification. In the present work, the O(2) and H(2)O(2) were separately combined with MgO used in cornstalk cooking for investigating their effects on delignification. After cooking, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service