Skip to Content
Merck
All Photos(1)

Key Documents

565423

Sigma-Aldrich

Acetonitrile solution

suitable for HPLC, contains 0.035 % (v/v) trifluoroacetic acid

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
4124158
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
81-82 °C
vapor pressure:
72.8 mmHg

Product Name

Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC

grade

HPLC grade

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg

form

liquid

contains

0.035 % (v/v) trifluoroacetic acid

expl. lim.

8 %

technique(s)

HPLC: suitable

bp

81-82 °C

mp

-48 °C

density

0.782 g/mL at 25 °C

application(s)

food and beverages

format

mixture

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is an acetonitrile solution containing 0.035%v/v trifluoroacetic acid. This pre-blended mobile phase solvent is of high purity that reduces time-consuming preparation and prevents instrument downtime because of impure mobile phases. Trifluoroacetic acid/acetonitrile mobile phase system is generally employed for peptide separation. Acetonitrile (MeCN), an organic solvent has low viscosity, high elution strength and is UV transparent at low wavelength. Trifluoroacetic acid (TFA) is commonly used as a mobile phase additive. It is a volatile acid that forms hydrophobic negatively charged trifluoroacetate ion.

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimum concentration of trifluoroacetic acid for reversed-phase liquid chromatography of peptides revisited.
Chen Y, et al.
Journal of Chromatography A, 1043(1), 9-18 (2004)
Eagleson M.
Concise Encyclopedia Chemistry, 6-6 (1994)
Acetonitrile, the polarity chameleon.
Zarzycki PK, et al.
Analytical and Bioanalytical Chemistry, 397(3), 905-908 (2010)
Hélène Hall et al.
Brain : a journal of neurology, 137(Pt 9), 2493-2508 (2014-07-27)
The neuropathological substrate of dementia in patients with Parkinson's disease is still under debate, particularly in patients with insufficient alternate neuropathology for other degenerative dementias. In patients with pure Lewy body Parkinson's disease, previous post-mortem studies have shown that dopaminergic
Elke H J Krekels et al.
Clinical pharmacokinetics, 53(6), 553-563 (2014-02-06)
From a previously validated paediatric population pharmacokinetic model, it was derived that non-linear morphine maintenance doses of 5 μg/kg(1.5)/h, with a 50 % dose reduction in neonates with a postnatal age (PNA) <10 days, yield similar morphine and metabolite concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service