Skip to Content
Merck
All Photos(1)

Key Documents

31250

Sigma-Aldrich

Potassium chromate

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CrO4
CAS Number:
Molecular Weight:
194.19
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Agency

reag. Ph. Eur.

vapor density

6.7 (vs air)

Assay

≥99.5%

impurities

≤0.005% in water insoluble matters

pH

8.6-9.8 (20 °C, 5%)

mp

971 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Fe: ≤20 mg/kg
Na: ≤200 mg/kg
Pb: ≤50 mg/kg

SMILES string

[K+].[K+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.2K.4O/q;2*+1;;;2*-1

InChI key

XMXNVYPJWBTAHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium chromate (K2CrO4) is the potassium salt having morphological features similar to potassium sulfate (isostructural). Its crystals exhibit rhombic crystal system. It can be manufactured by reacting potassium dichromate with potassium hydroxide.[1] It has been reported to crystallize in orthorhombic space group.[2] Production of potassium chromat by a new reaction system of KOH-KNO3-H2O (binary submolten salt system) has been proposed.[3]

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Redetermination of the structures of potassium sulphate and potassium chromate: the effect of electrostatic crystal forces upon observed bond lengths.
McGinnety JA.
Acta Crystallographica Section B, Structural Science, 28(9), 2845-2852 (1972)
Eagleson M.
Concise Encyclopedia Chemistry, 881-881 (1994)
Comparative Study of Factors Influencing the Action of Corrosion Inhmitors for Mild Steel in Neutral Solution: II. Potassium Chromate.
Mercer AD and Jenkins IR.
British Corrosion Journal, 3(3), 130-135 (1968)
A new method of potassium chromate production from chromite and KOH-KNO3-H2O binary submolten salt system.
Sun Z, et al.
AIChE Journal, 55(10), 2646-2656 (2009)
Subhendra N Mattagajasingh et al.
Journal of applied toxicology : JAT, 28(8), 987-997 (2008-07-11)
Hexavalent chromium [Cr(VI)] compounds are Group-I human carcinogens. Cr(VI)-induced DNA-protein crosslinks (DPCs) have been implicated in the mutagenic and carcinogenic effects of Cr(VI). Although multiple mechanisms have been suggested for Cr(VI)-induced DNA-protein crosslinking, the mechanism of formation of DNA-protein crosslinks

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service