Skip to Content
Merck
All Photos(1)

Key Documents

227870

Sigma-Aldrich

Nitroethane

ReagentPlus®, 99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2NO2
CAS Number:
Molecular Weight:
75.07
Beilstein:
1209324
EC Number:
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
grade:
reagent
bp:
114-115 °C (lit.)
vapor pressure:
15.6 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

reagent

vapor density

2.58 (vs air)

vapor pressure

15.6 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99.5%

form

liquid

autoignition temp.

778 °F

expl. lim.

3.4 %

dilution

(for general lab use)

refractive index

n20/D 1.391 (lit.)

bp

114-115 °C (lit.)

mp

−90 °C (lit.)

solubility

acetone: soluble(lit.)
alcohol: soluble(lit.)
water: slightly soluble(lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

CC[N+]([O-])=O

InChI

1S/C2H5NO2/c1-2-3(4)5/h2H2,1H3

InChI key

MCSAJNNLRCFZED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin Francis et al.
Biochemistry, 47(35), 9136-9144 (2008-08-12)
The deprotonation of nitroethane catalyzed by Neurospora crassa 2-nitropropane dioxygenase was investigated by measuring the formation and release of ethylnitronate formed in turnover as a function of pH and through mutagenesis studies. Progress curves for the enzymatic reaction obtained by
Paul F Fitzpatrick et al.
Archives of biochemistry and biophysics, 433(1), 157-165 (2004-12-08)
While several flavoproteins will oxidize nitroalkanes in addition to their physiological substrates, nitroalkane oxidase (NAO) is the only one which does not require the anionic nitroalkane. This, in addition to the induction of NAO by nitroethane seen in Fusarium oxysporum
S T Stokes et al.
The Journal of chemical physics, 129(6), 064308-064308 (2008-08-22)
Valence and dipole-bound negative ions of the nitroethane (NE) molecule and its clusters are studied using photoelectron spectroscopy (PES), Rydberg electron transfer (RET) techniques, and ab initio methods. Valence adiabatic electron affinities (EA(a)s) of NE, C(2)H(5)NO(2), and its clusters, (C(2)H(5)NO(2))(n)
Feng-Wu Liu et al.
Carbohydrate research, 344(18), 2439-2443 (2009-11-03)
Henry reactions of a novel higher sugar derivative, (1R)-(1,4:3,6-dianhydro-D-mannitol-2-yl)-1,4:3,6-dianhydro-D-fructose 5,5'-dinitrate (Alternate nomenclature: (1R)-(isomannid-2-yl)-1,4:3,6-dianhydro-D-fructose 5,5'-dinitrate), with nitromethane and nitroethane were studied. The kinetic and thermodynamic reactions with nitromethane under different conditions were carried out to afford (2S)- and (2R)-beta-nitroalcohols, respectively. But
Min Zhang et al.
Organic letters, 15(7), 1718-1721 (2013-03-27)
The Pd-catalyzed multidehydrogenative cross-coupling reactions of arenes with nitroethane are described. The established methods afford β-aryl nitroethylenes that are an important class of synthetic intermediates and biologically active compounds in an atom- and step-economical fashion. The reactions were applicable to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service