Skip to Content
Merck
All Photos(2)

Key Documents

221082

Sigma-Aldrich

Mercury(II) oxide yellow

ACS reagent, ≥99.0%

Synonym(s):

Mercuric oxide yellow

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgO
CAS Number:
Molecular Weight:
216.59
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
powder
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

≥99.0%

form

powder

reaction suitability

reagent type: catalyst
core: mercury

impurities

≤0.005% N compounds
≤0.03% insol. dil. HCl

reduction residue

≤0.05%

mp

500 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.025%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤0.003%

SMILES string

O=[Hg]

InChI

1S/Hg.O

InChI key

UKWHYYKOEPRTIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Yellow mercuric oxide shows antibacterial property and is commonly used in ophthalmic ointments.[1][2]

Application

Yellow mercuric oxide in the presence of mercuric bromide catalyst may be used in the preparation of alkyl β-D-glucopyranoside tetra-O-acetates and tetra-O-benzoates from poly-O-acyl-α-D-glucopyranosyl bromides and different alcohols via Koenigs-Knorr syntheses.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concise Inorganic Pharmaceutical Chemistry (phar.Che-I) (2008)
Koenigs-Knorr syntheses with mercuric salts.
Schroeder LR and Green JW.
J. Chem. Soc. Sect. C, 530-531 (1966)
T Endo et al.
Pharmacology & toxicology, 67(5), 431-435 (1990-11-01)
To obtain evidence for difference in the absorptive forms of HgCl2 and HgO from duodenum, the Hg distributions after the duodenal administration of HgCl2 and HgO and the effect of Na2SeO3 on their Hg distributions were compared. The Hg concentrations
Damir Krznarić et al.
Analytical chemistry, 80(3), 742-749 (2008-01-11)
Mercury electrodes preconcentrate metal chalcogenide nanoparticles effectively, enabling their detection at submicromolar concentrations (as Sigma chalcogenide) by adsorptive cathodic stripping voltammetry. Understanding the unique behavior of nanoparticle analytes during preconcentration is critical for lowering detection limits and for quantification. A
Inhalational mercury poisoning masquerading as toxic shock syndrome.
S B Mohan et al.
Anaesthesia and intensive care, 22(3), 305-306 (1994-06-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service