Skip to Content
Merck
All Photos(1)

Key Documents

11720

Sigma-Aldrich

Cadmium sulfate 8/3-hydrate

puriss., ≥98% (complexometric)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSO4 · 8/3H2O
CAS Number:
Molecular Weight:
256.57
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

puriss.

Assay

≥98% (complexometric)

form

solid

reaction suitability

reagent type: catalyst
core: cadmium

anion traces

chloride (Cl-): ≤50 mg/kg
nitrate (NO3-): ≤100 mg/kg

cation traces

As: ≤5 mg/kg
Fe: ≤10 mg/kg
Zn: ≤100 mg/kg

SMILES string

[Cd++].[Cd++].[Cd++].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/3Cd.3H2O4S.8H2O/c;;;3*1-5(2,3)4;;;;;;;;/h;;;3*(H2,1,2,3,4);8*1H2/q3*+2;;;;;;;;;;;/p-6

InChI key

AGHQMAQBLOTWPQ-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Target Organs

Bone,Kidney,Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hybrid CdSe/TiO2 nanowire photoelectrodes: Fabrication and photoelectric performance.
Ai g, et al.
Journal of Materials Chemistry, 21(24), 8749-8755 (2011)
Recovery of heavy metals from metal industry waste waters by chemical precipitation and nanofiltration.
Gonzalez-Munoz MJ, et al.
Desalination, 200(1), 742-744 (2006)
Syntheses, IR spectra, thermal properties and crystal structures of novel cyano-bridged polymeric complexes of zinc II and cadmium II with tetracyanoplatinate II.
Karadag A, et al.
Polyhedron, 27(1), 223-231 (2008)
Nicholas A T Irwin et al.
Current biology : CB, 31(1), 66-76 (2020-10-31)
DNA replication is a ubiquitous and conserved cellular process. However, regulation of DNA replication is only understood in a small fraction of organisms that poorly represent the diversity of genetic systems in nature. Here we used computational and experimental approaches
Els Keunen et al.
Plant physiology and biochemistry : PPB, 49(10), 1084-1091 (2011-08-20)
Plants exposed to cadmium (Cd) show morphological and physiological disorders. To increase our knowledge regarding Cd-induced signalling, most often the effects of acute exposure are investigated. However, this does not allow in-depth analysis of morphological effects. Therefore, we chronically exposed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service