Key Documents
Z803103
Specac® Atlas 15T manual hydraulic press
Select a Size
1 EA
R$49,092.00
R$49,092.00
Available to ship onApril 09, 2025Details
Request a Bulk Order
Select a Size
Change View
1 EA
R$49,092.00
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
R$49,092.00
Available to ship onApril 09, 2025Details
Request a Bulk Order
manufacturer/tradename
Specac Ltd
technique(s)
IR spectroscopy: suitable
General description
Specac® Atlas 15T manual hydraulic press may be used for the fabrication of free-standing bio-composites by employing two bio-polyesters with different elastic moduli and reinforced with different graphene nanoplatelets.[1] It may also be used to compress iron oxide granules to iron oxide discs to calculate the contact angle for the adsorption of heavy metals from wastewater using granulated iron oxide.[2]
Features and Benefits
  • Polycarbonate safety guards
  • Adjustable upper bolster
  • Adjustable pressure control valve
  • Vacuum ports
  • Pressure release valve
Legal Information
Specac is a registered trademark of Specac Ltd.
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Effect of graphene nano-platelet morphology on the elastic modulus of soft and hard biopolymers.
Cataldi P, et al.
Carbon, 109, 331-339 (2016)
Design, production and characterisation ofgranular adsorbent material for arsenic removalfrom contaminated wastewater.
Mangwadi C, et al.
Chemical Engineering Research and Design, 110, 70-81 (2016)