Y0002023
Plastic additive 26
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Trioctyl trimellitate, Tris(2-ethylhexyl) trimellitate
About This Item
grade
pharmaceutical primary standard
manufacturer/tradename
EDQM
refractive index
n20/D 1.485 (lit.)
bp
414 °C (lit.)
density
0.99 g/mL at 25 °C (lit.)
format
neat
storage temp.
2-8°C
SMILES string
CCCCC(CC)COC(=O)c1ccc(C(=O)OCC(CC)CCCC)c(c1)C(=O)OCC(CC)CCCC
InChI
1S/C33H54O6/c1-7-13-16-25(10-4)22-37-31(34)28-19-20-29(32(35)38-23-26(11-5)17-14-8-2)30(21-28)33(36)39-24-27(12-6)18-15-9-3/h19-21,25-27H,7-18,22-24H2,1-6H3
InChI key
KRADHMIOFJQKEZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Packaging
Other Notes
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
435.2 °F - closed cup
Flash Point(C)
224 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service