Skip to Content
Merck
All Photos(1)

Key Documents

Y0001161

Carbasalate impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-[[2-(Acetyloxy)benzoyl]oxy]benzoic acid, 2-Carboxyphenyl o-acetylsalicylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O6
CAS Number:
Molecular Weight:
300.26
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

carbasalate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O(c1c(cccc1)C(=O)Oc2c(cccc2)C(=O)O)C(=O)C

InChI

1S/C16H12O6/c1-10(17)21-14-9-5-3-7-12(14)16(20)22-13-8-4-2-6-11(13)15(18)19/h2-9H,1H3,(H,18,19)

InChI key

DDSFKIFGAPZBSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carbasalate impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F E Blondino et al.
Journal of pharmaceutical and biomedical analysis, 13(2), 111-119 (1995-02-01)
Formulation of pressurized aerosol solutions in propellants for inhalation requires the use of high quantities of surfactants to solubilize the drug. Due to the lipophilic nature of these surfactants, analytical difficulties are created for those wishing to quantify the drug
P V Mroso et al.
Journal of pharmaceutical sciences, 71(10), 1096-1101 (1982-10-01)
Salicylsalicylic acid and acetylsalicylsalicylic acid were identified as decomposition products of aspirin when mixtures of the drug, with magnesium stearate, were stored in the solid state of 60 degrees and 75% relative humidity. The effect of increasing the concentration of
R D Kirchhoefer et al.
Journal of pharmaceutical sciences, 69(5), 550-553 (1980-05-01)
A quantitative high-pressure liquid chromatographic method, using a reversed-phase column and an aqueous acetic acid-methanol solution as the mobile phase, was employed for the determination of O-acetyl-O-salicylsalicylic acid and O-salicylsalicylic acid in pharmaceutical aspirin preparations. The aspirin was dissolved, filtered
Tamás Fischer
Orvosi hetilap, 150(11), 503-512 (2009-03-05)
The beneficial effect achieved by the treatment of endothelial dysfunction in chronic cardiovascular diseases is already an evidence belonging to the basic treatment of the disease. Given the fact that the vascular system is uniform and consubstantial both physiologically, pathophysiologically
Hydrolysis of acetylsalicylsalicylic acid and salicylsalicylic acid in aqueous solution.
H Sunada et al.
Chemical & pharmaceutical bulletin, 33(5), 2158-2161 (1985-05-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service