Skip to Content
Merck
All Photos(1)

Key Documents

X4251

Supelco

XTT sodium salt

analytical standard, ≥90%

Synonym(s):

2,3-Bis(2-methoxy-4-nitro-5-sulfophenyl)-2H-tetrazolium-5-carboxanilide inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16N7NaO13S2
CAS Number:
Molecular Weight:
673.52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].COc1cc(c(cc1-n2nc(n[n+]2-c3cc(c(cc3OC)[N+]([O-])=O)S([O-])(=O)=O)C(=O)Nc4ccccc4)S([O-])(=O)=O)[N+]([O-])=O

InChI

1S/C22H17N7O13S2.Na/c1-41-17-8-15(28(31)32)19(43(35,36)37)10-13(17)26-24-21(22(30)23-12-6-4-3-5-7-12)25-27(26)14-11-20(44(38,39)40)16(29(33)34)9-18(14)42-2;/h3-11H,1-2H3,(H2-,23,30,35,36,37,38,39,40);/q;+1/p-1

InChI key

JACYMBNQPPWQML-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Useful in AIDS and cancer testing.
XTT has been used in conjunction with phenazine methosulfate (PMS) to screen human cancer cell lines. The tetrazolium dye has also been used to study fungal cell damage, in testing antimicrobial susceptibility of staphylococci, and in Candida biofilm research. Limitations to the XTT assay have been reported.

Principle

In living cells, XTT is metabolically reduced to produce a colorimetric, water-soluble formazan product.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parsons, J.L., et al.
Journal of Heterocyclic Chemistry, 25, 911-911 (1988)
Cristina Sánchez-Quesada et al.
Evidence-based complementary and alternative medicine : eCAM, 2018, 9473094-9473094 (2018-10-27)
Anti-inflammatory effects of virgin olive oil (VOO) have been described recently, along with its wound healing effect. One of the main minor compounds found in VOO is squalene (SQ), which also possesses preventive effects against skin damage and anti-inflammatory properties.
Wander José da Silva et al.
Brazilian dental journal, 19(4), 364-369 (2009-01-31)
2, 3-bis (2-methoxy-4-nitro-5-sulfophenyl)-5-[(phenylamino) carbonyl]-2H-tetrazolium hydroxide (XTT) reduction assay has been used to study Candida biofilm formation. However, considering that the XTT reduction assay is dependent on cell activity, its use for evaluating mature biofilms may lead to inaccuracies since biofilm
D A Scudiero et al.
Cancer research, 48(17), 4827-4833 (1988-09-01)
We have previously described the application of an automated microculture tetrazolium assay (MTA) involving dimethyl sulfoxide solubilization of cellular-generated 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)-formazan to the in vitro assessment of drug effects on cell growth (M.C. Alley et al., Proc. Am. Assoc.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service