Skip to Content
Merck
All Photos(1)

Documents

W417920

Sigma-Aldrich

2-Methylfuran

99%

Synonym(s):

Silvan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O
CAS Number:
Molecular Weight:
82.10
FEMA Number:
4179
Beilstein:
103733
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 117

vapor density

2.8 (vs air)

vapor pressure

139 mmHg ( 20 °C)
561 mmHg ( 55 °C)

Assay

99%

refractive index

n20/D 1.433 (lit.)

bp

63-66 °C (lit.)

density

0.91 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

chocolate; ethereal

SMILES string

Cc1ccco1

InChI

1S/C5H6O/c1-5-3-2-4-6-5/h2-4H,1H3

InChI key

VQKFNUFAXTZWDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methylfuran (2-MF) is a 2-alkyl substituted furan. Its microwave spectrum has been analyzed. The synthesis of 2-MF from carbon sources under different reaction conditions have been reported.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of λ-butyrolactone and 2-methylfuran through the coupling of dehydrogenation and hydrogenation over copper-chromite catalyst.
Zheng HY, et al.
Reaction Kinetics and Catalysis Letters, 82(2), 263-269 (2004)
Efficient production of the liquid fuel 2,5-dimethylfuran from fructose using formic acid as a reagent.
Todsapon Thananatthanachon et al.
Angewandte Chemie (International ed. in English), 49(37), 6616-6618 (2010-08-04)
Ke Xiong et al.
ChemSusChem, 7(8), 2146-2149 (2014-04-24)
Selectively cleaving the C=O bond outside the furan ring of furfural is crucial for converting this important biomass-derived molecule to value-added fuels such as 2-methylfuran. In this work, a combination of density functional theory (DFT) calculations, surface science studies, and
Jinglei Cui et al.
ChemSusChem, 9(11), 1259-1262 (2016-04-28)
An efficient process was designed for the synthesis of furfuryl alcohol and 2-methylfuran from xylose using a continuous fixed-bed reactor over a catalyst combining Hβ zeolite and Cu/ZnO/Al2 O3 in γ-butyrolactone (GBL)/water as solvent. The cooperative effect of Hβ zeolite
Microwave Spectrum of 2-Methylfuran.
Niorrs WG and Krisher LC.
J. Chem. Phys. , 51, 403-406 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service