Skip to Content
Merck
All Photos(1)

Key Documents

T62200

Sigma-Aldrich

Trifluoroacetic acid

ReagentPlus®, 99%

Synonym(s):

TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
742035
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

reagent

vapor density

3.9 (vs air)

vapor pressure

97.5 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

refractive index

n20/D 1.3 (lit.)

bp

72.4 °C (lit.)

mp

−15.4 °C (lit.)

solubility

H2O: soluble

density

1.489 g/mL at 20 °C (lit.)
1.478 g/mL at 25 °C (exp. value)

SMILES string

OC(C(F)(F)F)=O

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>212.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 100 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukiko Enomoto-Rogers et al.
Carbohydrate polymers, 92(2), 1827-1834 (2013-02-13)
Fully substituted glucomannan triacetate (GMTAc) (degree of substitution (DS)=3.0) was prepared from konjac glucomannan (KGM) treated with acetic acid and trifluoroacetic anhydride (TFAA). The peaks in the (1)H- and (13)C NMR spectra of GMTAc were assigned in detail based on
Guillaume Plourde et al.
Lancet (London, England), 386(10009), 2192-2203 (2015-09-29)
Transradial access for cardiac catheterisation results in lower bleeding and vascular complications than the traditional transfemoral access route. However, the increased radiation exposure potentially associated with transradial access is a possible drawback of this method. Whether transradial access is associated
Julia Kennedy-Darling et al.
Journal of proteome research, 13(8), 3810-3825 (2014-07-08)
DNA-protein interactions play critical roles in the control of genome expression and other fundamental processes. An essential element in understanding how these systems function is to identify their molecular components. We present here a novel strategy, Hybridization Capture of Chromatin
Nichollas E Scott et al.
Molecular & cellular proteomics : MCP, 13(9), 2354-2370 (2014-06-12)
The opportunistic human pathogen Acinetobacter baumannii is a concern to health care systems worldwide because of its persistence in clinical settings and the growing frequency of multiple drug resistant infections. To combat this threat, it is necessary to understand factors
Caroline Le Maréchal et al.
Veterinary research, 42, 35-35 (2011-02-18)
Staphylococcus aureus is a major cause of mastitis in ruminants. In ewe mastitis, symptoms range from subclinical to gangrenous mastitis. S. aureus factors or host-factors contributing to the different outcomes are not completely elucidated. In this study, experimental mastitis was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service