Skip to Content
Merck
All Photos(1)

Documents

T2005000

Triflusal

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

α,α,α-trifluoro-2,4-cresotic acid acetate, 2-(Acetyloxy)-4-(trifluoromethyl)benzoic acid, Drisgen, acetyl-4-trifluoromethylsalicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7F3O4
CAS Number:
Molecular Weight:
248.16
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

triflusal

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C10H7F3O4/c1-5(14)17-8-4-6(10(11,12)13)2-3-7(8)9(15)16/h2-4H,1H3,(H,15,16)

InChI key

RMWVZGDJPAKBDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Triflusal EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Triflusal is an anti-inflammatory antithrombotic platelet aggregation inhibitor with a 3-fold mechanism of action. It is an irreversible inhibitor of COX-1 inhibiting thromboxane A2, preventing aggregation; it preserves vascular prostacyclin, thus promoting anti-aggregant effect. Triflusal blocks phosphodiesterase leading to an increase in cAMP levels promoting anti-aggregant effect due to inhibition of calcium mobilization.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Murdoch et al.
Drugs, 66(5), 671-692 (2006-04-20)
Triflusal (Aflen, Disgren, Tecnosal, Triflux) is a novel platelet antiaggregant with structural similarities to salicylates, but which is not derived from aspirin. It has similar efficacy to aspirin in patients with cerebral or myocardial infarction, but has a reduced risk
José Antonio González-Correa et al.
Cardiovascular drug reviews, 24(1), 11-24 (2006-08-31)
Triflusal is a derivative of salicylic acid with a well-established platelet aggregation inhibitory profile. Its pharmacokinetic and pharmacodynamic properties differ, however, somewhat from those of acetylsalicylic acid. A number of recent experimental and clinical studies have shown that triflusal is
Triflusal: an old drug in modern antiplatelet therapy. Review of its action, use, safety and effectiveness.
Hector Anninos et al.
Hellenic journal of cardiology : HJC = Hellenike kardiologike epitheorese, 50(3), 199-207 (2009-05-26)
J Alvarez Sabín et al.
Revista de neurologia, 27(160), 951-955 (1999-02-10)
Different studies have shown that aspirin (AAS), in low doses, may lead to a considerable frequency of hemorrhagic complications when used in the long term. We compare the long-term occurrence of hemorrhagic complications with low doses of AAS and high
J Costa et al.
The Cochrane database of systematic reviews, (3)(3), CD004296-CD004296 (2005-07-22)
Aspirin is the standard treatment for secondary prevention of stroke and other vascular events. Several studies suggest that triflusal may have a better safety profile. To determine in people at high risk of vascular events whether triflusal is an effective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service