Skip to Content
Merck
All Photos(5)

Documents

SRE0096

Sigma-Aldrich

Bovine Serum Albumin

≥98.0%, New Zealand Origin, ≤3 EU/mg endotoxin, protein quantification

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41141801

product name

Bovine Serum Albumin, heat shock fraction, protease free, IgG free, low endotoxin, Ph 7.0, ≥98%

grade

standard grade

Assay

≥98.0%

form

powder

mol wt

~66 kDa

purified by

heat shock fractionation

origin

New Zealand origin

availability

available only in China

technique(s)

protein quantification: suitable

impurities

≤3 EU/mg endotoxin

pH

7.0

cation traces

heavy metals (as Pb): ≤10 ppm

foreign activity

Ribonuclease, none detected
endonuclease, none detected
protease ≤0.005 units/mg solid
proteinase, none detected

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Preparation Note

Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunzhi Liu et al.
International journal of molecular medicine, 48(4) (2021-08-21)
Omega‑3 polyunsaturated fatty acids (n‑3 PUFAs) exert a negative effect on IL‑6 production in several liver disorders, including cirrhosis, acute liver failure and fatty liver disease. However, its effect on the production of IL‑11, another important IL‑6 family cytokine, remains
Zitong Huang et al.
Journal of neuroscience methods, 348, 109012-109012 (2020-11-30)
The nerve plexus of the enteric nervous system (ENS) plays a crucial part in regulating gastrointestinal functions, such as muscle contractile activity and nutrient absorption. Studying the nerve plexus can provide vital information for research on ENS disorders. Whole-mount preparation
PACAP attenuates hepatic lipid accumulation through the FAIM/AMPK/IRI? axis during overnutrition.
Luo, et al.
Molecular Metabolism, 65, 101584-101584 (2022)
Ni Wang et al.
Journal of ginseng research, 47(3), 448-457 (2023-05-30)
Alzheimer's disease (AD) is a common form of dementia, and impaired mitophagy is a hallmark of AD. Mitophagy is mitochondrial-specific autophagy. Ginsenosides from Ginseng involve in autophagy in cancer. Ginsenoside Rg1 (Rg1 hereafter), a single compound of Ginseng, has neuroprotective
Erling Hu et al.
iScience, 25(3), 103966-103966 (2022-03-15)
The bioeffects of terahertz (THz) radiation received growing attention because of its influence on the interactions between biomolecules. Our work aimed to investigate the effects of THz irradiation on cell membrane, especially cell membrane permeability. We found that 0.1 THz

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service