Skip to Content
Merck
All Photos(1)

Documents

SMB00959

Sigma-Aldrich

trans-Resveratrol 3-O-sulfate potassium salt

Synonym(s):

Potassium 3-hydroxy-5-[(1E)-2-(4-hydroxyphenyl)ethenyl]phenyl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11KO6S
CAS Number:
Molecular Weight:
346.40
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

synthetic

Quality Level

form

solid

technique(s)

HPLC: suitable

color

white to beige

storage temp.

2-8°C

Application

Metabolomics research
Resveratrol-3-O-sulfate is a metabolite derived from trans-Resveratrol. It can be found in biological fluids. Within the cell, Resveratrol 3-sulfate is primarily located in the cytoplasm and in the extracellular space

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service