Skip to Content
Merck
All Photos(1)

Key Documents

S5263

Sigma-Aldrich

Sulfanilic acid

≥99.0%

Synonym(s):

4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(H2N)C6H4SO3H
CAS Number:
Molecular Weight:
173.19
Beilstein:
908765
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99.0%

mp

>300 °C (lit.)

SMILES string

Nc1ccc(cc1)S(O)(=O)=O

InChI

1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

HVBSAKJJOYLTQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nam Hoon Kim et al.
Nano letters, 10(10), 4181-4185 (2010-09-25)
A bifunctional adenosine-sensitive double-stranded DNA aptamer was used to create and control a surface-enhanced Raman spectroscopy (SERS) hot spot between a bulk Au surface and a gold nanoparticle (Au NP) attached to the aptamer via a biotin-avidin linkage. The Au
Brian D Piorek et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(48), 18898-18901 (2007-11-21)
We present a microfluidic technique for sensitive, real-time, optimized detection of airborne water-soluble molecules by surface-enhanced Raman spectroscopy (SERS). The method is based on a free-surface fluidic device in which a pressure-driven liquid microchannel flow is constrained by surface tension.
Reversible tuning of SERS hot spots with aptamers.
Nam Hoon Kim et al.
Advanced materials (Deerfield Beach, Fla.), 23(36), 4152-4156 (2011-08-09)
Kwan Kim et al.
Langmuir : the ACS journal of surfaces and colloids, 27(8), 4526-4531 (2011-03-17)
4-Aminobenzenthiol (4-ABT) is an unusual molecule, showing variable surface-enhanced Raman scattering (SERS) spectra depending upon measurement conditions. In an effort to reduce ambiguity and add clarity, we have thus conducted an ultraviolet-visible (UV-vis) extinction measurement, along with Raman scattering measurement
Ke-Jing Huang et al.
Colloids and surfaces. B, Biointerfaces, 61(2), 176-181 (2007-09-18)
A chemically modified electrode is constructed based on the multi-walled carbon nanotubes (MWNTs)/4-aminobenzeresulfonic acid (4-ABSA) film-coated glassy carbon electrode. The electrocatalytic oxidation of tyrosine (Tyr) is investigated on the surface of the MWNTs/4-ABSA-modified electrode using cyclic voltammetry (CV) and differential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service