PHR8948
6-epi-Doxycycline HCl
certified reference material, pharmaceutical secondary standard
Synonym(s):
2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, 6-epimer, monohydrochloride.
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
API family
doxycycline
packaging
pkg of 50MG
application(s)
pharmaceutical
storage temp.
-10 to -25°C
General description
The Oligonucleotide HPLC Performance Standard Mix (Oligo-6) is a calibration standard with carefully selected oligonucleotides to test and monitor changes in HPLC and LC/MS Systems. The difference in the selectivity of the oligonucleotides make it useful for most chromatographic modes including Ion Pairing Reversed Phase, Ion Exchange and HILIC Methods.
The oligonucleotides in the mix are dried and between 12-33 NT that makes it a suitable tool alongside development and quality control of therapeutics like antisense oligonucleotides, small interfering RNAs, and aptamers.
Components
Sequence 1, 12 NT, 3588.4 Da
Sequence 2, 12 NT, 4158.0 Da
Sequence 3, 25 NT, 7580.9 Da
Sequence 4, 33 NT, 10014.4 Da
Sequence 5, 20 NT, 6117.0 Da
Sequence 6, 15 NT, 4395.9 Da
Application
- System suitability and creation of control charts
- Method transfer and troubleshooting
- Monitoring of HPLC Column life span IPRP-HPLC, IEX HPLC, HILIC, LCMS.
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service