Skip to Content
Merck
All Photos(1)

Key Documents

PHR2611

Supelco

Rifabutin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Rifabutin, Ansamycin, Ansatipine (Farmitalia), LM-427, Mycobutin (Farmitalia)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$1,934.00

R$1,934.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
R$1,934.00

About This Item

Empirical Formula (Hill Notation):
C46H62N4O11
CAS Number:
Molecular Weight:
847.00
UNSPSC Code:
41116107
NACRES:
NA.24

R$1,934.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000149
traceable to USP 1603800

description

Pharmaceutical Secondary Standard; Certified Reference Material

API family

rifabutin

form

solid

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

application(s)

pharmaceutical small molecule

storage temp.

-10 to -25°C

SMILES string

[N+H]1(CCC2(NC3=C4NC(=O)\C(=C/C=C/[C@@H]([C@@H]([C@H]([C@H]([C@H]([C@@H]([C@@H]([C@H](\C=C\O[C@]5(Oc6c(c(c(c(c6C)[O-])C4=O)C3=N2)C5=O)C)OC)C)OC(=O)C)C)O)C)O)C)\C)CC1)CC(C)C

InChI

1S/C46H62N4O11/c1-22(2)21-50-18-16-46(17-19-50)48-34-31-32-39(54)28(8)42-33(31)43(56)45(10,61-42)59-20-15-30(58-11)25(5)41(60-29(9)51)27(7)38(53)26(6)37(52)23(3)13-12-14-24(4)44(57)47-36(40(32)55)35(34)49-46/h12-15,20,22-23,25-27,30,37-38,41,49,52-54H,16-19,21H2,1-11H3,(H,47,57)/b13-12+,20-15+,24-14-/t23-,25+,26+,27+,30-,37-,38+,41+,45-/m0/s1

InChI key

ATEBXHFBFRCZMA-VXTBVIBXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Rifabutin is an antibiotic; antitumor.
Rifabutin is an antibiotic; antitumor. Rifabutin interferes with HSP-90 molecular chaperone, enhances ubiquitination and protein degradation, and inactivates bacterial RNA polymerase.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service