Skip to Content
Merck
All Photos(1)

Key Documents

PHR2272

4-Dehydroxy-4-chloro Penciclovir

new

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14ClN5O2
CAS Number:
Molecular Weight:
271.70
UNSPSC Code:
12352200

Pricing and availability is not currently available.

grade

certified reference material

Quality Level

API family

penciclovir

packaging

pkg of 100MG

application(s)

pharmaceutical

storage temp.

2-8°C

Other Notes

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service