Skip to Content
Merck
All Photos(1)

Key Documents

PHR2137

Supelco

Ibuprofen Methyl Ester

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ibuprofen Methyl Ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$1,739.00

R$1,739.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
R$1,739.00

About This Item

Empirical Formula (Hill Notation):
C14H20O2
CAS Number:
Molecular Weight:
220.31
UNSPSC Code:
41116107
NACRES:
NA.24

R$1,739.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

ibuprofen

form

liquid

CofA

current certificate can be downloaded

packaging

pkg of 1 mL

application(s)

pharmaceutical small molecule

storage temp.

2-30°C

SMILES string

O(C)C(=O)C(C)c1ccc(cc1)CC(C)C

InChI

1S/C14H20O2/c1-10(2)9-12-5-7-13(8-6-12)11(3)14(15)16-4/h5-8,10-11H,9H2,1-4H3

InChI key

YNZYUHPFNYBBFF-UHFFFAOYSA-N

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service