PHR1918
Carbamazepine Related Compound A
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
10,11-Dihydrocarbamazepine, 10,11-Dihydro-5H-dibenz[b,f]azepine-5-carboxamide
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to Ph. Eur. Y0000033
traceable to USP 1093012
API family
carbamazepine
form
powder
CofA
current certificate can be downloaded
packaging
pkg of 50 mg
mp
205-210 °C (lit.)
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
NC(=O)N1c2ccccc2CCc3ccccc13
InChI
1S/C15H14N2O/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-8H,9-10H2,(H2,16,18)
InChI key
PHNLCHMJDSSPDQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Repr. 1A - Skin Sens. 1A - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service