Skip to Content
Merck
All Photos(1)

Documents

PHR1722

Supelco

Triamterene

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Triamterene, 2,4,7-Triamino-6-phenylpteridine, 6-Phenyl-2,4,7-pteridinetriamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N7
CAS Number:
Molecular Weight:
253.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000837
traceable to USP 1680007

API family

triamterene

CofA

current certificate can be downloaded

packaging

pkg of 400 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Nc1nc(N)c2nc(-c3ccccc3)c(N)nc2n1

InChI

1S/C12H11N7/c13-9-7(6-4-2-1-3-5-6)16-8-10(14)18-12(15)19-11(8)17-9/h1-5H,(H6,13,14,15,17,18,19)

InChI key

FNYLWPVRPXGIIP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Triamterene is a potassium-sparing diuretic, which exhibits antihypertensive activity. It is used in combination with thiazide deuretic, such as benzthiazide for treating hypertension and edema associated with mild to moderate congestive heart failure.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Triamterene may be used as a secondary pharma standard for the quantification of the analyte in pharmaceutical formulations using reversed-phase high-performance liquid chromatography technique.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7397 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and Validation of RP-HPLC Method for Simultaneous Estimation of Triamterene and Benzthiazide in Tablets
Chauhan CV, et al.
PharmaTutor, 2(6), 115-122 (2014)
High performance liquid chromatography for simultaneous determination of xipamide, triamterene and hydrochlorothiazide in bulk drug samples and dosage forms
El-Hay ASS, et al.
Acta pharmaceutica, 66(1), 109-118 (2016)
Xinyu Qi et al.
Nature medicine, 25(8), 1225-1233 (2019-07-25)
Polycystic ovary syndrome (PCOS) is characterized by androgen excess, ovulatory dysfunction and polycystic ovaries1, and is often accompanied by insulin resistance2. The mechanism of ovulatory dysfunction and insulin resistance in PCOS remains elusive, thus limiting the development of therapeutics. Improved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service