Skip to Content
Merck
All Photos(1)

Key Documents

PHR1519

Supelco

Sulfacetamide Sodium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Sulfacetamide sodium salt monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$427.00

R$427.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$427.00

About This Item

Empirical Formula (Hill Notation):
C8H9N2NaO3S · H2O
CAS Number:
Molecular Weight:
254.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$427.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. S1700000
traceable to USP 1624006

API family

sulfacetamide

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC1=CC=C(S(N([Na])C(C)=O)(=O)=O)C=C1.O

InChI

1S/C8H10N2O3S.Na.H2O/c1-6(11)10-14(12,13)8-4-2-7(9)3-5-8;;/h2-5H,9H2,1H3,(H,10,11);;1H2/q;+1;/p-1

InChI key

IHCDKJZZFOUARO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sulfacetamide Sodium belongs to the class of sulfonamides,[1] widely used in ophthalmic preparations, to treat different ocular infections such as corneal ulcers and conjunctivitis. It shows bactericidal activity against susceptible microorganisms.[2]

Application

Sulfacetamide Sodium may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations[1][3] by spectrometric technique[1] and micellar electro-kinetic chromatography.[3]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3326 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of prednisolone acetate, sulfacetamide and phenylefrine in local pharmaceutical preparations by micellar electrokinetic chromatography
Gallego LMJ and Arroyo PJ
Journal of Pharmaceutical and Biomedical Analysis, 31(5), 873-884 (2003)
Validation of a stability-indicating spectrometric method for the determination of sulfacetamide sodium in pure form and ophthalmic preparations
Ahmed S, et al.
Journal of Pharmacy & Bioallied Sciences, 9(2), 126-126 (2017)
Pharmacology, 9(2), 126-126 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service