Skip to Content
Merck
All Photos(1)

Documents

PHR1468

Supelco

Quinic Acid

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

D-(−)-Quinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O6
CAS Number:
Molecular Weight:
192.17
Beilstein:
2212412
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1594506

API family

quinic acid

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

O[C@@H]1C[C@@](O)(C[C@@H](O)[C@H]1O)C(O)=O

InChI

1S/C7H12O6/c8-3-1-7(13,6(11)12)2-4(9)5(3)10/h3-5,8-10,13H,1-2H2,(H,11,12)/t3-,4-,5-,7+/m1/s1

InChI key

AAWZDTNXLSGCEK-WYWMIBKRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Quinic Acid is a plant metabolite and a chiral building block used in multistep chemical synthesis of natural compounds.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3016 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aditya Arya et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 71, 183-196 (2014-06-24)
The aim of this study was to investigate the synergistic effects of quercetin (QE) and quinic acid (QA) on a STZ-induced diabetic rat model to determine their potential role in alleviating diabetes and its associated complications. In our study design
Diana M Cheng et al.
Nutrition (Burbank, Los Angeles County, Calif.), 30(7-8 Suppl), S52-S58 (2014-07-06)
The aims of the following experiments were to characterize antidiabetic in vitro and in vivo activity of the polyphenol-rich aqueous extract of Rutgers Scarlet Lettuce (RSL). RSL extract (RSLE) and isolated compounds were evaluated for inhibitory effects on glucose production as well
Mengqi Ye et al.
Journal of the science of food and agriculture, 94(14), 2951-2957 (2014-03-13)
Polyphenols and organic acids are important constitutes in the cider because they greatly contribute to organoleptic quality. The determination of their changes is important for monitoring the fermentation process for purposes of quality control. In this study, the evolution of
Hong-Fei Wang et al.
Antonie van Leeuwenhoek, 107(3), 835-843 (2015-01-09)
A white bacterial strain, designated EGI 650022(T), was isolated from the roots of Salsola affinis C. A. Mey, collected from Urumqi City, Xinjiang, north-western China. The strain was found to be aerobic, Gram-stain positive, oxidase-positive and catalase-positive. Cells were non-motile
Xin-Hua Lu et al.
Molecules (Basel, Switzerland), 19(6), 8518-8532 (2014-06-25)
The fruit physico-chemical properties, antioxidant activity and mineral contents of 26 pineapple [Ananas comosus (L.) Merr.] genotypes grown in China were measured. The results showed great quantitative differences in the composition of these pineapple genotypes. Sucrose was the dominant sugar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service