Skip to Content
Merck
All Photos(1)

Key Documents

PHR1403

Supelco

Phentolamine Mesylate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Phentolamine methanesulfonate salt, 2-(N-[m-Hydroxyphenyl]-p-toluidinomethyl)imidazoline

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$518.00

R$518.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$518.00

About This Item

Linear Formula:
C17H19N3O · CH3SO3H
CAS Number:
Molecular Weight:
377.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$518.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001127
traceable to USP 1530004

API family

phentolamine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

180-182 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CS(O)(=O)=O.Cc1ccc(cc1)N(CC2=NCCN2)c3cccc(O)c3

InChI

1S/C17H19N3O.CH4O3S/c1-13-5-7-14(8-6-13)20(12-17-18-9-10-19-17)15-3-2-4-16(21)11-15;1-5(2,3)4/h2-8,11,21H,9-10,12H2,1H3,(H,18,19);1H3,(H,2,3,4)

InChI key

OGIYDFVHFQEFKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phentolamine Mesylate belongs to the class of alpha adrenergic antagonist drugs. It is used in the treatment of pheochromocytoma-related hypertension, erectile dysfunction and in the diagnosis of pheochromocytoma and norepinephrine-related dermal necrosis.[1][2][3]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Phentolamine Mesylate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chemiluminescence technique.[4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

α-Adrenergic receptor antagonist; peripheral vasodilator.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3261 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacokinetics of lidocaine with epinephrine following local anesthesia reversal with phentolamine mesylate
Moore PA, et al.
Anesthesia Progress, 55(2), 40-48 (2008)
Phentolamine mesylate relaxes penile corpus cavernosum tissue by adrenergic and non-adrenergic mechanisms
Traish A, et al.
International Journal of Impotence Research, 10(4), 215-215 (1998)
Sensitive chemiluminescence determination of phentolamine mesylate and phenoxybenzamine hydrochloride based on K3Fe (CN) 6-H2O2-fluorescein
Xing L, et al.
Journal of Luminescence, 137(4), 162-167 (2013)
Long-term safety and efficacy of oral phentolamine mesylate (Vasomax?) in men with mild to moderate erectile dysfunction
Padma-Nathan H, et al.
International Journal of Impotence Research, 14(4), 266-266 (2002)
Xianming Su et al.
Journal of cardiovascular pharmacology, 65(1), 62-71 (2014-09-30)
Previous studies have found that Danhong injection (DHI), an extensively used herbal extract preparation in China, might be a powerful vasodilator. The aims of this study were to determine the vascular activity of DHI and its effects on arteries of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service