Skip to Content
Merck
All Photos(1)

Key Documents

PHL89905

Tomatine

phyproof® Reference Substance

Synonym(s):

Lycopersicin

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$2,408.00

R$2,408.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
R$2,408.00

About This Item

Empirical Formula (Hill Notation):
C50H83NO21
CAS Number:
Molecular Weight:
1034.19
Beilstein:
78250
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$2,408.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

mp

300-305 °C (dec.)

application(s)

food and beverages

format

neat

SMILES string

C[C@H]1CCC2(NC1)O[C@H]3C[C@H]4[C@@H]5CC[C@H]6C[C@H](CC[C@]6(C)[C@H]5CC[C@]4(C)[C@H]3[C@@H]2C)O[C@@H]7O[C@H](CO)[C@H](O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O[C@@H]9OC[C@@H](O)[C@H](O)[C@H]9O)[C@H]8O[C@@H]%10O[C@H](CO)[C@@H](O)[C@H](O)[C@H]%10O)[C@H](O)[C@H]7O

InChI

1S/C50H83NO21/c1-20-7-12-50(51-15-20)21(2)32-28(72-50)14-26-24-6-5-22-13-23(8-10-48(22,3)25(24)9-11-49(26,32)4)65-45-40(63)37(60)41(31(18-54)68-45)69-47-43(71-46-39(62)36(59)34(57)29(16-52)66-46)42(35(58)30(17-53)67-47)70-44-38(61)33(56)27(55)19-64-44/h20-47,51-63H,5-19H2,1-4H3/t20-,21-,22-,23-,24+,25-,26-,27+,28-,29+,30+,31+,32-,33-,34+,35+,36-,37+,38+,39+,40+,41-,42-,43+,44-,45+,46-,47-,48-,49-,50-/m0/s1

InChI key

REJLGAUYTKNVJM-SGXCCWNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simona Serratì et al.
International journal of molecular sciences, 21(15) (2020-07-29)
There is a growing interest in the cytotoxic effects of bioactive glycoalkaloids, such as α-tomatine on tumor cells. Here, for the first time, we determine the antitumor potential of tomatine, a mixture of α-tomatine and dehydrotomatine, in metastatic melanoma (MM)
Elenn Suzany Pereira Aranha et al.
Toxicology in vitro : an international journal published in association with BIBRA, 66, 104879-104879 (2020-05-04)
Melanoma is a skin cancer with high invasive potential and high lethality. Considering that quinonemethide triterpenes are described as promising anticancer agents, the aim of this study was to evaluate the effect of 22β-hydroxytingenone (22-HTG) against human melanoma cells. Alamar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service