Skip to Content
Merck
All Photos(1)

Key Documents

PHL80379

Cardamomin

phyproof® Reference Substance

Synonym(s):

Cardamonin, (2E)-1-(2,4-Dihydroxy-6-methoxyphenyl)-3-phenyl-2-propen-1-one, (8CI); Alpinetin chalcone, (E)-2′,4′-Dihydroxy-6′-methoxy-chalcone

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$3,177.00

R$3,177.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
R$3,177.00

About This Item

Empirical Formula (Hill Notation):
C16H14O4
CAS Number:
Molecular Weight:
270.28
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$3,177.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

storage temp.

2-8°C

SMILES string

O=C(/C=C/C1=CC=CC=C1)C2=C(O)C=C(O)C=C2OC

InChI

1S/C16H14O4/c1-20-15-10-12(17)9-14(19)16(15)13(18)8-7-11-5-3-2-4-6-11/h2-10,17,19H,1H3/b8-7+

InChI key

NYSZJNUIVUBQMM-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Cardamonin is a calchone from Aplinia species (zingiberaceous plant species).
Cardamonin is a calchone from Aplinia species (zingiberaceous plant species). Cardamonin inhibits pigmentation in melanocytes through suppression of Wnt/b-catenin signaling pathway. Cardamonin suppressed Wnt/b-catenin signaling by a mechanism involving proteasome-mediated degradation of b-catenin (GSK-3b-independent pathway). Research has shown that Cardamonin possesses anti-inflammatory activity via suppression of NF-kB nuclear translocation and Ik-Ba phosphorylation.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service