Skip to Content
Merck
All Photos(1)

Key Documents

P9352

Millipore

Perfringens T.S.C. supplement

suitable for microbiology

Synonym(s):

Perfringens Tryptose-Sulfite-Cycloserine-Supplement, Tryptose Sulphite Cycloserine Supplement, Perfringens

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
R$684.00

R$684.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 VIAL
R$684.00

About This Item

EC Number:
UNSPSC Code:
41171614
NACRES:
NA.85

R$684.00


Check Cart for Availability

Request more information

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Clostridium spp.

Application

Recommended for the selective isolation of Clostridium perfringens.

Components

Component: D-Cycloserine 200.00 mg

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service