Skip to Content
Merck
All Photos(1)

Key Documents

P9146

Sigma-Aldrich

Sodium pyrophosphate tetrabasic decahydrate

≥98% (titration)

Synonym(s):

Pyrophosphate inorganic, Sodium diphosphate tetrabasic, Tetrasodium pyrophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na4P2O7 · 10H2O
CAS Number:
Molecular Weight:
446.06
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥98% (titration)

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

InChI

1S/4Na.H4O7P2.10H2O/c;;;;1-8(2,3)7-9(4,5)6;;;;;;;;;;/h;;;;(H2,1,2,3)(H2,4,5,6);10*1H2/q4*+1;;;;;;;;;;;/p-4

InChI key

VZWGHDYJGOMEKT-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taís Scaramucci et al.
Journal of dentistry, 39(12), 841-848 (2011-09-29)
To evaluate the erosive potential of orange juice modified by food additives in enamel and dentine. Calcium lactate pentahydrate (CLP), xanthan gum (XG), sodium linear polyphosphate (LPP), sodium pyrophosphate tetrabasic (PP), sodium tripolyphosphate (STP) and some of their combinations were
Priyadip Das et al.
Chemical communications (Cambridge, England), 47(28), 8118-8120 (2011-06-21)
Newly synthesized Zn(II) and Cd(II)-based complexes show unique selectivity towards inorganic pyrophosphate (PPi) in 100% aqueous medium at pH = 7.4 and act as a "turn-on" and "turn-off" real-time assay, respectively, for the evaluation of the enzymatic activity of alkaline
Nadia Marino et al.
Inorganic chemistry, 50(1), 378-389 (2010-12-01)
The reaction in water of Cu(NO(3))(2)·2.5H(2)O with 2,2'-bipyridine (bipy), 1,10-phenanthroline (phen), or 1,10-phenanthroline-5-amine (phenam), and sodium pyrophosphate (Na(4)P(2)O(7)), at various pHs, afforded three new copper(II)-pyrophosphate complexes, namely, {[Cu(bipy)(cis-H(2)P(2)O(7))](2)}·3H(2)O (1a), {[Cu(phen)(H(2)O)](4)(HP(2)O(7))(2)}(ClO(4))(2)·4H(2)O (2), and {[Cu(2)(phenam)(2)(P(2)O(7))](2)·25H(2)O}(n) (3). A solvent free crystalline phase of
Bruce L Riser et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 26(10), 3349-3357 (2011-03-15)
The high rate of cardiovascular mortality in patients with end-stage renal disease (ESRD) is a significant barrier to improved life expectancy. Unique in this population is the marked development and aggressive worsening of vascular calcification (VC). Pyrophosphate (PPi), an endogenous
Xingqiang Wu et al.
Analytica chimica acta, 709, 66-72 (2011-11-30)
The optimization of analytical procedures for the quantification of free and total microcystins (MCs) in natural sediments was systematically examined based on solvent extraction and Lemieux oxidation. In this optimized analytical procedure, a sequential solvent extraction using 50% (v/v) methanol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service