Skip to Content
Merck
All Photos(1)

Key Documents

P5221

Sigma-Aldrich

2x Acculong PCR ReadyMix

suitable for PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55
Pricing and availability is not currently available.

form

liquid

Quality Level

technique(s)

PCR: suitable

shipped in

dry ice

storage temp.

−20°C

General description

2x Acculong PCR ReadyMix contains Acculong DNA Polymerase, dNTP and an optimized buffer system, only requiring the addition of primers and templates to perform amplification.
This reduces the need for pipetting operations, improves detection throughput, and yields reproducible results. In addition, the protective agent added in the system allows the 2x Acculong PCR ReadyMix to maintain stable activity after repeated freezing and thawing.

Features and Benefits

2x Acculong PCR ReadyMix has the following advantages:


  • High Fidelity :53 times that of Taq DNA Polymerase and 6 times that of Pfu DNA Polymerase.
  • Faster Amplification:Limit speed can reach 0.5 sec/kb, 30 sec/kb can efficiently amplify most fragments


Longer Amplification:
  • Effective amplification length of simple templates can reach 40 kb.
  • Effective amplification length of genome can reach 20 kb.
  • Effective amplification length of cDNA can reach 10 kb.


Widely Applicable:
  • Suitable for amplification of various GC content fragments.
  • Super tolerance to PCR inhibitors.
  • Can be used for direct PCR of a variety of samples.
  • Easy-to-Use:Dye premix minimizing experimental steps

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Utilize molecular raw materials for diagnostic assay development and manufacturing, backed by extensive experience and worldwide manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service