Skip to Content
Merck
All Photos(1)

Key Documents

NIST927F

Bovine Serum Albumin (7% Solution)

NIST® SRM® 927f

Synonym(s):

BSA 7% solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Product NIST927F is currently not available for purchase. Contact Technical Service

grade

certified reference material

Agency

NIST

packaging

pack of 10 × 2.2 mL each

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable
protein quantification: suitable

application(s)

clinical

format

single component solution

storage temp.

2-8°C

General description

This bovine serum albumin solution (BSA) is a standard reference material (SRM) of known protein concentration (approximately 7%) and purity. The NIST-certified value is determined with Amino Acid Analysis using ID-LC/MS/MS. In addition, a reference value using the Biuret method is provided. NIST927F consists of 10 ampoules, each containing approximately 2.2 mL of solution.

NIST927F_Cert

NIST927F_SDS

Application

This albumin solution from bovine serum is used:
  • for standardization of procedures employed in clinical analyses for total serum protein
  • for critical evaluation of daily working standards used in these procedures
  • as a reference standard for assays of total protein by colorimetric methods
  • for gel diffusion, amino acid analysis, electrophoresis, HPLC, and nitrogen assays
  • for other tests that require well-characterized protein for calibration or evaluation

Other Notes

  • Refer to the certificate for expiry, maintenance, storage, usage, and preparation of dilutions details.
  • Notes for biomaterials, disposal, and transport information are available in SDS.
Certified for the analytes listed below.
Albumin

See certificate for values and more details at nist.gov/SRM.

Warning

SRM 927f IS INTENDED FOR RESEARCH USE.

Storage and Stability

This SRM is supplied to the user in sealed ampoules. The SRM should be stored in a refrigerator at a temperature between 2 °C and 8 °C. The ampoules should not be frozen because of possible breakage of ampoules during the thawing process.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service